Learn More
Aviva Systems Biology Corp ALOX15B Antibody - C-terminal region (ARP41895_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41895_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ALOX15B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ALOX15B. Alternate names: 15-LOX-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58038