Learn More
Aviva Systems Biology Corp ALOX15B Antibody - N-terminal region (ARP41894_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41894_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALOX15B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ALOX15B. Alternate names: 15-LOX-2ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58037