Learn More
TARGETMOL CHEMICALS INC ALOXISTATIN 10MG
SDP
Catalog No. 502576619
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Aloxistatin (E64d) is an inhibitor of cysteine protease with blood platelet aggregation inhibiting activity. Aloxistatin is an irreversible membrane-permeable inhibitor of lysosomal and cytosolic cysteine proteases with the ability to inhibit calpain activity in intact platelets. purity: 99%
Catalog No. 50-257-6619 Supplier TARGETMOL CHEMICALS INC Supplier No. T604010MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Aloxistatin (E64d) is an inhibitor of cysteine protease with blood platelet aggregation inhibiting activity. Aloxistatin is an irreversible membrane-permeable inhibitor of lysosomal and cytosolic cysteine proteases with the ability to inhibit calpain activity in intact platelets. purity: 99%