Learn More
Medchemexpress LLC HY-100229 10mg Medchemexpress, Aloxistatin CAS:88321-09-9 Purity:>98%
SDP
Catalog No. 501967717
Encompass_Preferred
Medchemexpress, HY-100229 10mg Aloxistatin CAS:88321-09-9 Aloxistatin (E64d) is a cell-permeable and irreversible broad-spectrum cysteine protease inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7717 Supplier Medchemexpress LLC Supplier No. HY10022910MG
Medchemexpress, HY-100229 10mg Aloxistatin CAS:88321-09-9 Aloxistatin (E64d) is a cell-permeable and irreversible broad-spectrum cysteine protease inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.