Learn More
Medchemexpress LLC HY-100229 25mg , Aloxistatin E64d;E64c ethyl ester CAS:88321-09-9 Purity:98%
SDP
Catalog No. 501493995
Encompass_Preferred
Medchemexpress, HY-100229 25mg Aloxistatin E64d;E64c ethyl ester CAS:88321-09-9 Formula:C17H30N2O5 Cysteine protease Purity:98% Aloxistatin (E64d) is a broad-spectrum cell-permeable cysteine protease inhibitor. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3995 Supplier Medchemexpress LLC Supplier No. HY10022925MG
Medchemexpress, HY-100229 25mg Aloxistatin E64d;E64c ethyl ester CAS:88321-09-9 Formula:C17H30N2O5 Cysteine protease Purity:98% Aloxistatin (E64d) is a broad-spectrum cell-permeable cysteine protease inhibitor. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.