Learn More
Beckman Coulter B87478, Automated Lab Positioner
Catalog No. NC1890825
Encompass_Preferred
The Beckman Coulter B87478 is an automated lab positioner for high-throughput screening in drug discovery. It has a 3-axis robotic arm for precise labware positioning and a compact design to save space. Compatible with various lab devices, it supports microplates, tube racks, and more. Its software enables flexible programming and third-party integration, enhancing workflow automation in genomics and drug research.
Catalog No. NC1890825 Supplier Beckman Coulter Supplier No. B87478
The Beckman Coulter B87478 is an automated lab positioner for high-throughput screening in drug discovery. It has a 3-axis robotic arm for precise labware positioning and a compact design to save space. Compatible with various lab devices, it supports microplates, tube racks, and more. Its software enables flexible programming and third-party integration, enhancing workflow automation in genomics and drug research.