Learn More
Cayman Chemical ALP Assay Buffer, 30 mL
SDP
Catalog No. 502945261
Encompass_Preferred
ALP Assay Buffer has been tested and formulated to work exclusively with Cayman’s Alkaline Phosphatase Colorimetric Activity Assay Kit (Item No. 701710).
Catalog No. 50-294-5261 Supplier Cayman Chemical Supplier No. 40008330 ML
ALP Assay Buffer has been tested and formulated to work exclusively with Cayman’s Alkaline Phosphatase Colorimetric Activity Assay Kit (Item No. 701710).