Learn More
Rockland Immunochemicals ALP Conjugate Stabilizer (Azide and Mercury Free) - MB-061-0100
SDP
Catalog No. 501059018
Encompass_Preferred
ALP Conjugate Stabilizer is to be used with calf intestinal Alkaline Phosphatase (ALP) conjugated primary or secondary antibodies or other proteins or compounds in solution.   The components of the stabilizer (proprietary) are unique, and stabilize both the antibody and enzyme portions of the conjugate. ALP Conjugate Stabilizer prevents the loss of catalytic activity and maintains the active conformation of the antibody portion of the conjugate.
Catalog No. 50-105-9018 Supplier Rockland Immunochemicals Supplier No. MB0610100
ALP Conjugate Stabilizer is to be used with calf intestinal Alkaline Phosphatase (ALP) conjugated primary or secondary antibodies or other proteins or compounds in solution.   The components of the stabilizer (proprietary) are unique, and stabilize both the antibody and enzyme portions of the conjugate. ALP Conjugate Stabilizer prevents the loss of catalytic activity and maintains the active conformation of the antibody portion of the conjugate.