Learn More
Diagnostic Biosystems ALP LABEL Conjugated Enzyme Labels
Catalog No. 502407429
Encompass_Preferred
Diagnostic BioSystems offers immunostaining detection systems based on streptavidin/biotin technology. We offer species-specific kits to etect antibodies raised in rabbits or mice and convenient broad spectrum kits to detect both. DBS detection systems are available in ready-touse formats, with the secondary antibody containing either goat anti-mouse and anti-rabbit IgG, goat anti-mouse IgG, or goat anti-rabbit IgG.
Catalog No. 50-240-7429 Supplier Diagnostic Biosystems Supplier No. M010
Add to Cart
Edge
Add to Cart
Diagnostic BioSystems offers immunostaining detection systems based on streptavidin/biotin technology. We offer species-specific kits to etect antibodies raised in rabbits or mice and convenient broad spectrum kits to detect both. DBS detection systems are available in ready-touse formats, with the secondary antibody containing either goat anti-mouse and anti-rabbit IgG, goat anti-mouse IgG, or goat anti-rabbit IgG.