Learn More
Cayman Chemical ALP Substrate 50mg
SDP
Catalog No. 502942784
Encompass_Preferred
ALP Substrate has been tested and formulated to work exclusively with Cayman’s Alkaline Phosphatase Colorimetric Activity Assay Kit (Item No. 701710).
Catalog No. 50-294-2784 Supplier Cayman Chemical Supplier No. 40011250 MG
ALP Substrate has been tested and formulated to work exclusively with Cayman’s Alkaline Phosphatase Colorimetric Activity Assay Kit (Item No. 701710).