Learn More
Beckman Coulter ALP Trash, Coulter
Catalog No. NC1890826
Encompass_Preferred
The Beckman Coulter ALP Trash is an automated pipetting system for high-throughput applications. It has an 8-channel arm, tool changer, and supports 96/384 well plates. With 4 channels, it handles 0.5-300 μL volumes using D.A.R.T. technology. Ideal for PCR, plate replication, and dilutions, it saves benchtop space and includes software for method management.
Catalog No. NC1890826 Supplier Beckman Coulter Supplier No. B87483
The Beckman Coulter ALP Trash is an automated pipetting system for high-throughput applications. It has an 8-channel arm, tool changer, and supports 96/384 well plates. With 4 channels, it handles 0.5-300 μL volumes using D.A.R.T. technology. Ideal for PCR, plate replication, and dilutions, it saves benchtop space and includes software for method management.