Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alpelisib | 1217486-61-7 | C19H23ClF3N5O2S | 5 MG
SDP

Catalog No. 5000373658
Encompass_Preferred

Alpelisib hydrochloride is a potent, orally active, and selective PI3Kα inhibitor with IC50s of 5 nM for p110α. It exhibits antineoplastic activity and potently inhibits Akt phosphorylation in cells transformed with PI3Kα.

  • Potently inhibits PIK3CA somatic mutations (H1047R, E545K; IC50s~4 nM).
  • Significantly reduces Akt phosphorylation in cells transformed with PI3Kα (IC50=74±15 nM).
  • Inhibits cell growth of osteosarcoma cell lines.
  • Induces G0/G1 phase cell cycle arrest in human and murine osteosarcoma cells.
  • Reduces tumor volumes and deposition of ectopic bone matrix in animal models.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 2 years or -20°C for 1 year.
  • Shipping can occur at room temperature if duration is less than 2 weeks.

Catalog No. 50-003-73658 Supplier Medchemexpress LLC Supplier No. HY15244A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alpelisib hydrochloride is a potent, orally active, and selective PI3Kα inhibitor with IC50s of 5 nM for p110α. It exhibits antineoplastic activity and potently inhibits Akt phosphorylation in cells transformed with PI3Kα.

  • Potently inhibits PIK3CA somatic mutations (H1047R, E545K; IC50s~4 nM).
  • Significantly reduces Akt phosphorylation in cells transformed with PI3Kα (IC50=74±15 nM).
  • Inhibits cell growth of osteosarcoma cell lines.
  • Induces G0/G1 phase cell cycle arrest in human and murine osteosarcoma cells.
  • Reduces tumor volumes and deposition of ectopic bone matrix in animal models.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 2 years or -20°C for 1 year.
  • Shipping can occur at room temperature if duration is less than 2 weeks.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.