Learn More
Medchemexpress LLC HY-15244 500mg , Alpelisib CAS:1217486-61-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15244500MG
Encompass_Preferred
HY-15244 500mg Alpelisib CAS: 1217486-61-7 Alpelisib (BYL-719) is a potent, selective, and orally active PI3Kα inhibitor. Alpelisib (BYL-719) shows efficacy in targeting PIK3CA-mutated cancer. Alpelisib (BYL-719) also inhibits p110α/p110γ/p110δ/p110β with IC50s of 5/250/290/1200 nM, respectively. Antineoplastic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-8995