Learn More
Medchemexpress LLC Alpelisib hydrochloride | 1584128-91-5 | 99.6% | 477.93 g/mol | C19H23ClF3N5O2S | 10 MG
SDP
Catalog No. 5000286603
Encompass_Preferred
Alpelisib hydrochloride is the hydrochloride salt of a selective PI3Kα inhibitor used in research to probe PI3K signaling and PIK3CA-driven oncogenic pathways. Supplied as a high-purity solid in small laboratory pack sizes, it is intended for in vitro and preclinical studies; handle per safety data and institutional protocols.
  • Selective PI3Kα inhibitor for pathway and cancer research.
  • High purity (≈99.6%) suitable for analytical and biological assays.
  • Light yellow to yellow solid physical form.
  • Available in small research pack sizes, including 10 mg.
  • Includes core identifiers: CAS number, formula, and molecular weight.
  • Store sealed at 4°C; in solvent, store at -80°C.
Catalog No. 50-002-86603 Supplier Medchemexpress LLC Supplier No. HY15244A10MG
Alpelisib hydrochloride is the hydrochloride salt of a selective PI3Kα inhibitor used in research to probe PI3K signaling and PIK3CA-driven oncogenic pathways. Supplied as a high-purity solid in small laboratory pack sizes, it is intended for in vitro and preclinical studies; handle per safety data and institutional protocols.
  • Selective PI3Kα inhibitor for pathway and cancer research.
  • High purity (≈99.6%) suitable for analytical and biological assays.
  • Light yellow to yellow solid physical form.
  • Available in small research pack sizes, including 10 mg.
  • Includes core identifiers: CAS number, formula, and molecular weight.
  • Store sealed at 4°C; in solvent, store at -80°C.