Learn More
Medchemexpress LLC Alpelisib hydrochloride | 1584128-91-5 | C19H23ClF3N5O2S | 500 MG
SDP
Catalog No. 5000372270
Encompass_Preferred
Alpelisib hydrochloride (BYL-719) is a potent, selective, and orally active PI3Kα inhibitor with efficacy in targeting PIK3CA-mutated cancer. It exhibits antineoplastic activity.
  • Potent, orally active, and selective PI3Kα inhibitor
  • Inhibits PIK3CA somatic mutations (H1047R, E545K)
  • Inhibits Akt phosphorylation in PI3Kα transformed cells
  • Induces G0/G1 phase cell cycle arrest in osteosarcoma cell lines
  • Significantly reduces tumor volumes in animal models
Catalog No. 50-003-72270 Supplier Medchemexpress LLC Supplier No. HY15244A500MG
Alpelisib hydrochloride (BYL-719) is a potent, selective, and orally active PI3Kα inhibitor with efficacy in targeting PIK3CA-mutated cancer. It exhibits antineoplastic activity.
  • Potent, orally active, and selective PI3Kα inhibitor
  • Inhibits PIK3CA somatic mutations (H1047R, E545K)
  • Inhibits Akt phosphorylation in PI3Kα transformed cells
  • Induces G0/G1 phase cell cycle arrest in osteosarcoma cell lines
  • Significantly reduces tumor volumes in animal models