Learn More
BEST SANITIZERS Alpet D2 Quat-Free Surface Sanitizer, 5 gal for use with ADB0001, ADB0002
Catalog No. NC1498182
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Best Sanitizers' Footwear Sanitizing Stations add an additional layer of pathogen protection by reducing cross-contamination from footwear. Utilizing the proven efficacy of Alpet D2 Quat-Free Surface Sanitizer, facilities now have a powerful tool for controlling moisture, pathogens and cross-contamination with one of our footwear sanitizing stations (ADB0001/ADB0002). Ready-to-Use, no-rinse required 70% Alcohol Formula (62.5% ethanol / 7.5% isopropanol) NSF listed, Kosher, Pareve and Halal certified Highly Evaporative and ideal for low moisture environments and water sensitive equipment Kills 99.9% of tested pathogens in 10 seconds on non-food contact surfaces, including rubber footwear Approved under the Washington State Department of Agriculture Organic Food Program
Catalog No. NC1498182 Supplier BEST SANITIZERS Supplier No. SS10031BSS
This item has a minimum qty of 2 per supplier requirements.
Best Sanitizers' Footwear Sanitizing Stations add an additional layer of pathogen protection by reducing cross-contamination from footwear. Utilizing the proven efficacy of Alpet D2 Quat-Free Surface Sanitizer, facilities now have a powerful tool for controlling moisture, pathogens and cross-contamination with one of our footwear sanitizing stations (ADB0001/ADB0002). Ready-to-Use, no-rinse required 70% Alcohol Formula (62.5% ethanol / 7.5% isopropanol) NSF listed, Kosher, Pareve and Halal certified Highly Evaporative and ideal for low moisture environments and water sensitive equipment Kills 99.9% of tested pathogens in 10 seconds on non-food contact surfaces, including rubber footwear Approved under the Washington State Department of Agriculture Organic Food Program