Learn More
BEST SANITIZERS Alpet D2 Surface Sanitizer, 5 gal for use with ADB0001, ADB0002
Catalog No. NC1403160
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Best Sanitizers' Footwear Sanitizing Stations add an additional layer of pathogen protection by reducing cross-contamination from footwear . Utilizing the proven efficacy of Alpet D2 Surface Sanitizer, facilities now have a powerful tool for controlling moisture, pathogens and cross-contamination with one of our footwear sanitizing stations (ADB0001/ADB0002). Alpet D2 is a ready-to-use, no-rinse, fast acting, food contact and non-food contact surface cleaner and sanitizer. 58% IPA/quat formula Clean and Sanitize with one product 10 Second Kill on non-food contact surfaces, including rubber footwear** Kills 99.999% of 9 tested pathogens as a food contact surface sanitizer Highly Evaporative and ideal for water sensitive equipment and low moisture environments NSF Listed, EPA Registered, Kosher, Pareve and Halal**
This item is not returnable. View return policy
Catalog No. NC1403160 Supplier BEST SANITIZERS Supplier No. SS10002BSS
This item has a minimum qty of 2 per supplier requirements.
Best Sanitizers' Footwear Sanitizing Stations add an additional layer of pathogen protection by reducing cross-contamination from footwear . Utilizing the proven efficacy of Alpet D2 Surface Sanitizer, facilities now have a powerful tool for controlling moisture, pathogens and cross-contamination with one of our footwear sanitizing stations (ADB0001/ADB0002). Alpet D2 is a ready-to-use, no-rinse, fast acting, food contact and non-food contact surface cleaner and sanitizer. 58% IPA/quat formula Clean and Sanitize with one product 10 Second Kill on non-food contact surfaces, including rubber footwear** Kills 99.999% of 9 tested pathogens as a food contact surface sanitizer Highly Evaporative and ideal for water sensitive equipment and low moisture environments NSF Listed, EPA Registered, Kosher, Pareve and Halal**