Learn More
BEST SANITIZERS Alpet D2 Surface Sanitizing Wipes, 90 ct. Heavy Duty
Catalog No. NC1323950
Encompass_Preferred
This item has a minimum qty of 2 cases per supplier requirements.
Alpet D2 Surface Sanitizing Wipes are a ready-to-use, no-rinse, lintless wiping system for food contact surfaces. Alpet D2 Surface Sanitizing Wipes are available in a heavy duty 90 count wipe . As with all Best Sanitizers wiping systems, the spring-loaded lid closes on its own to prevent the drying of wipes, helping to eliminate unnecessary waste.Kills 99.999% of Staphylococcus aureus and Escherichia coli on hard, non-porous surfaces in 60 seconds. Strong, lintless wipes for food contact surfaces Spring-loaded lid to prevent drying Ready-to-use, no-rinse required 58% IPA/quat formula ideal for water sensitive equipment NSF listed D2 classification Kosher, Pareve and Halal certified
This item is not returnable. View return policy
Catalog No. NC1323950 Supplier BEST SANITIZERS Supplier No. SSW0001
This item has a minimum qty of 2 cases per supplier requirements.
Alpet D2 Surface Sanitizing Wipes are a ready-to-use, no-rinse, lintless wiping system for food contact surfaces. Alpet D2 Surface Sanitizing Wipes are available in a heavy duty 90 count wipe . As with all Best Sanitizers wiping systems, the spring-loaded lid closes on its own to prevent the drying of wipes, helping to eliminate unnecessary waste.Kills 99.999% of Staphylococcus aureus and Escherichia coli on hard, non-porous surfaces in 60 seconds. Strong, lintless wipes for food contact surfaces Spring-loaded lid to prevent drying Ready-to-use, no-rinse required 58% IPA/quat formula ideal for water sensitive equipment NSF listed D2 classification Kosher, Pareve and Halal certified