Learn More
New England Biolabs, Inc. α1-2,3 Mannosidase - 640 units
SDP
Catalog No. 50811846
Encompass
1-2,3 Mannosidase is a highly specific exoglycosidase that catalyzes the hydrolysis of 1-2 and 1-3 linked mannose residues from oligosaccharides
Catalog No. 50-811-846 Supplier New England Biolabs, Inc. Supplier No. P0729S
1-2,3 Mannosidase is a highly specific exoglycosidase that catalyzes the hydrolysis of 1-2 and 1-3 linked mannose residues from oligosaccharides