Learn More
New England Biolabs, Inc. α1-2 Fucosidase - 1000 units
SDP
Catalog No. 50811838
Encompass
a1-2 Fucosidase is a highly specific exoglycosidase that catalyzes the hydrolysis of linear 1-2 linked fucose residues from oligosaccharides
Catalog No. 50-811-838 Supplier New England Biolabs, Inc. Supplier No. P0724S
a1-2 Fucosidase is a highly specific exoglycosidase that catalyzes the hydrolysis of linear 1-2 linked fucose residues from oligosaccharides