Learn More
QA Bio Inc Alpha-(1-2,3,6)-Mannosidase - 60 µls
Catalog No. 502236629
Encompass
Alpha Mannosidase from Jack Bean cleaves α(1-2,3,6)-linked mannose. • This enzyme is often used in conjunction with Core Alpha-(1-6) Mannosidase (E-AM02) if a noncleavable core α(1-6) mannose is present on the substrate. • Source: Jack Bean • EC 3.2.1.24 • Contents: Alpha-(1-2,3,6) Mannosidase in 150 mM sodium phosphate, 0.1 mM ZnCl2 pH 7.5. (pH 7.5). / 200 µl 5x Reaction Buffer 5.0 (250 mM sodium phosphate, pH 5.0). • Specific Activity >3 U/mg • Activity >10 U/ml • Molecular weight two polypeptides of 64,000 and 44,000 daltons • Specificty All Alpha-(1-2,3,6)-linked mannose
This item is not returnable. View return policy
Catalog No. 50-223-6629 Supplier QA Bio Inc Supplier No. EAM01
Alpha Mannosidase from Jack Bean cleaves α(1-2,3,6)-linked mannose. • This enzyme is often used in conjunction with Core Alpha-(1-6) Mannosidase (E-AM02) if a noncleavable core α(1-6) mannose is present on the substrate. • Source: Jack Bean • EC 3.2.1.24 • Contents: Alpha-(1-2,3,6) Mannosidase in 150 mM sodium phosphate, 0.1 mM ZnCl2 pH 7.5. (pH 7.5). / 200 µl 5x Reaction Buffer 5.0 (250 mM sodium phosphate, pH 5.0). • Specific Activity >3 U/mg • Activity >10 U/ml • Molecular weight two polypeptides of 64,000 and 44,000 daltons • Specificty All Alpha-(1-2,3,6)-linked mannose