Learn More
New England Biolabs, Inc. α1-3,6 Galactosidase - 100 units
SDP
Catalog No. 508112030
Encompass
1-3,6 Galactosidase is a highly specific exoglycosidase that catalyzes the hydrolysis of 1-3 and 1-6 linked galactose residues from oligosaccharides.
Catalog No. 50-811-2030 Supplier New England Biolabs, Inc. Supplier No. P0731S
1-3,6 Galactosidase is a highly specific exoglycosidase that catalyzes the hydrolysis of 1-3 and 1-6 linked galactose residues from oligosaccharides.