Learn More
QA Bio Inc Alpha-(1-3,6)-Galactosidase
Supplier:  QA Bio Inc EAG02
Encompass
Alpha Galactosidase from E. coli cleaves α(1-3)- and α(1-6)-linked, non-reducing terminal galactose from complex carbohydrates and glycoproteins. There is no activity on α(1-4) linked galactose. It is particularly efficient for removing α-linked galactose under conditions where the pH must be neutral or above, for example, with living cells. • For cleaving Beta Galactosidase we recommend ß-(1-4) Galactosidase (E-BG07), recombinant from Streptococcus pneumoniae or ß-(1,3,4,6) Galactosidase (E-BG02), purified from bovine testes. • Source: recombinant from E. coli in E. coli • Contents: Alpha Galactosidase enzyme in 50 mM sodium phosphate, pH 7.5 / 5x Reaction Buffer – 250 mM sodium phosphate, pH 6.5 • Specific Activity >30 U/mg • Activity 80 U/ml • Molecular weight ~80,000 daltons • Specifictity: Cleaves Alpha-(1-3)-and Alpha-(1-6)-linked, non-reducing terminal galactose from complex carbohydrates and glycoproteins.
Catalog No. 50-223-6623