Learn More
New England Biolabs, Inc. α1-6 Mannosidase - 800 units
SDP
Catalog No. 50811842
Encompass
a1-6 Mannosidase is a highly specific exoglycosidase that removes unbranched a1-6 linked mannose residues from oligosaccharides. When used in conjunction with a1-2,3 Mannosidase, the a1-6 Mannosidase will cleave a1-6 mannose residues from branched carbohydrate substrates.
Catalog No. 50-811-842 Supplier New England Biolabs, Inc. Supplier No. P0727S
a1-6 Mannosidase is a highly specific exoglycosidase that removes unbranched a1-6 linked mannose residues from oligosaccharides. When used in conjunction with a1-2,3 Mannosidase, the a1-6 Mannosidase will cleave a1-6 mannose residues from branched carbohydrate substrates.