Learn More
Bioss Alpha-1-Acid Glycoprotein Mono
SDP
Catalog No. 5000067606
Encompass_Preferred
Alpha-1-Acid Glycoprotein Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IHC-P IHC-F IF RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-67606 Supplier Bioss Supplier No. BSM61556R
Alpha-1-Acid Glycoprotein Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IHC-P IHC-F IF RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol