Learn More
Alpco Ltd Canine Alpha 1-Antitrypsin ELISA
SDP
Supplier:  Alpco Ltd 41A1ACAE01
Encompass
The Alpha 1-Antitrypsin (A1AT) test kits are a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring A1AT in biological fluid of dogs. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-751-3567