Learn More
New England Biolabs, Inc. α2-3 Neuraminidase S - 400 units
SDP
Catalog No. 501047512
Encompass
a2-3 Neuraminidase S is a highly specific exoglycosidase that catalyzes the hydrolysis of a2-3 linked N-acetylneuraminic acid residues from oligosaccharides and glycoproteins.
Catalog No. 50-104-7512 Supplier New England Biolabs, Inc. Supplier No. P0743S
a2-3 Neuraminidase S is a highly specific exoglycosidase that catalyzes the hydrolysis of a2-3 linked N-acetylneuraminic acid residues from oligosaccharides and glycoproteins.