Learn More
Carbosynth LLC. Alpha-2-Gliadin (57-89), 5mg, MW: 3911.55, Molecular formula: C190H273N43O47
SDP
Supplier:  Carbosynth LLC. GIL3994PI5MG
Encompass_Preferred
Synonyms: H-Leu-Gln-Leu-Gln-Pro-Phe-Pro-Gln-Pro-Gln-Leu-Pro-Tyr-Pro-Gln-Pro-Gln-Leu-Pro-Tyr-Pro-Gln-Pro-Gln-Leu-Pro-Tyr-Pro-Gln-Pro-Gln-Pro-Phe-OH, α2-gliadin (57-89), α-gliadin 33-mer epitope, 33mer-alpha gliadin Peptide (57-89), H-LQLQPFPQPQLPYPQPQLPYPQPQLPYPQPQPF-OH, Application: Antigenic Determanent for Celiac Disease and a component of gluten, Storage Temperature: -20 °C, References: M. De Angelis, et al., Appl. Environ. Microbiol., 76 (2), 508 (2010)., K. Schalk, et al., Scientific Reports, 7, 45092 (2017)., B. Moron, Am J Clin Nutr, 87(2), 405 (2008).
Catalog No. 50-168-5782