Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-(4-Chlorophenyl)-2-pyridinemethanol 98.0+%, TCI America™
SDP

Catalog No. C26461G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26461G 1 g
C26465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26461G Supplier TCI America Supplier No. C26461G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27652-89-7
Molecular Formula C12H10ClNO
Molecular Weight (g/mol) 219.67
MDL Number MFCD00023493
InChI Key ZFUPOFQRQNJDNS-UHFFFAOYNA-N
PubChem CID 97719
IUPAC Name (4-chlorophenyl)(pyridin-2-yl)methanol
SMILES OC(C1=CC=C(Cl)C=C1)C1=CC=CC=N1

Specifications

Melting Point 84°C
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Formula Weight 219.67
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material alpha-(4-Chlorophenyl)-2-pyridinemethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.