Learn More
Cole-Parmer PCRmax Alpha Cycler 4 Thermal Cycler, Quad 96-Well Capacity; 100 to 230 VAC
Supplier:  Cole-Parmer 9394512
Encompass_Preferred
These compact thermal cyclers deliver reproducible results every runwith little to no training needed. Features include a clear responsive touch screen, secure user-specific programming, adjustable heated lid, and active sample cooling for sharper amplification. A Program Wizard generates a protocol specific to your sequence in secondsa quick way to optimize new assays. Units also retain approximately 1000 reports for reviewing at a later date and run on an intuitive, HD Android™ tablet interface. Save your personal settings to your USB drive; sign in is as simple as inserting your USB drive, no password needed.
Catalog No. NC1774365
Add to Cart
Request a Quote