Learn More
Enzo Life Sciences alpha Actinin 4 polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ALX210356C050
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 28-38 (M28GDYMAQEDDW38) of human and rat actinin-4 and to aa 29-39 of mouse actinin-4. Species reactivity: Human, Mouse, Rat Porcine, Chinese hamster. Creossreactivity: Does not cross-react with actinin-1, -2 or -3. Applications: IHC (FS), IHC (PS), IP, WB. Recommended Dilutions: Immunohistochemistry (frozen sections, 1µg/ml) Immunoprecipitation (25µg/ml) Western Blot (0.1-0.25µg/ml). The recommended dilutions refer to the analysis of mouse cells and tissues with intermediate levels of actinin-4 expression and must be viewed as approximate Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Epitope-affinity purified. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.01% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-8118