Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha,alpha'-Dihydroxy-1,3-diisopropylbenzene 98.0+%, TCI America™
SDP

Catalog No. D178825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D178825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D178825G Supplier TCI America Supplier No. D178825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1999-85-5
Molecular Formula C12H18O2
Molecular Weight (g/mol) 194.27
MDL Number MFCD00059139
InChI Key UGPWRRVOLLMHSC-UHFFFAOYSA-N
Synonym 1,3-Bis(alpha-hydroxyisopropyl)benzene, 2,2′C-(m-Phenylene)di-2-propanol, alpha,alpha,alpha′C,alpha′C-Tetramethyl-1,3-benzenedimethanol
PubChem CID 74816
IUPAC Name 2-[3-(2-hydroxypropan-2-yl)phenyl]propan-2-ol
SMILES CC(C)(O)C1=CC(=CC=C1)C(C)(C)O

Specifications

Melting Point 139°C
Color White
Quantity 25 g
Formula Weight 194.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha,alpha′-Dihydroxy-1,3-diisopropylbenzene
EINECSNumber (3)-3621
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.