Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha,alpha-Diphenyl-4-pyridylmethanol 98.0+%, TCI America™
SDP

Catalog No. D208925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D208925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D208925G Supplier TCI America Supplier No. D208925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1620-30-0
Molecular Formula C18H15NO
Molecular Weight (g/mol) 261.32
MDL Number MFCD00006434
InChI Key MRHLFZXYRABVOZ-UHFFFAOYSA-N
PubChem CID 74177
IUPAC Name diphenyl(pyridin-4-yl)methanol
SMILES OC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=NC=C1

Specifications

Melting Point 242°C
Color Red-White
Quantity 25 g
Formula Weight 261.32
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material alpha,alpha-Diphenyl-4-pyridylmethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.