Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha,alpha-Diphenyl-N-methyl-D-prolinol, 97%, ee 99+%, Thermo Scientific™

Catalog No. AAL19398MD
Change view
Click to view available options
Quantity:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19398MD 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL19398MD Supplier Thermo Scientific Chemicals Supplier No. L19398MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 144119-12-0
Quantity 250mg
Optical Rotation −52° (c=1 in Chloroform)
Molecular Formula C18H22NO
MDL Number MFCD00145217
Beilstein 1535407
Synonym r-1-methylpyrrolidin-2-yl diphenylmethanol, r---2-hydroxy diphenyl methyl-1-methylpyrrolidine, r-alpha,alpha-diphenylmethylprolinol, 2r-1-methylpyrrolidin-2-yl-diphenylmethanol, 1,1-diphenyl-n-methyl-d-prolinol, 2r-1-methylpyrrolidin-2-yl diphenylmethanol, r-n-methyl-alpha,alpha-diphenyl-2-pyrrolidinemethanol, r-a,a-diphenylmethylprolinol, d-diphenylmethylprolinol, r---diphenyl 1-methylpyrrolidin-2-yl methanol
InChI Key XIJAGFLYYNXCAB-QGZVFWFLSA-O
SMILES C[NH+]1CCC[C@@H]1C(O)(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name (2R)-2-(hydroxydiphenylmethyl)-1-methylpyrrolidin-1-ium
Molecular Weight (g/mol) 268.38
PubChem CID 853010
Formula Weight 267.37
Percent Purity 97%
Assay ee ≥99%
Chemical Name or Material alpha,alpha-Diphenyl-N-methyl-D-prolinol
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.