Learn More
Strem, An Ascensus Company CAS# 1344-28-1. 500g. α-Alumina trihydrate, 65% Al2O3, 34.8% H2O, 0.15% Na2O. MFCD00149140
SDP
Supplier:  Strem, An Ascensus Company 131000500G
Encompass
CAS# 1344-28-1. 500g. α-Alumina trihydrate, 65% Al2O3, 34.8% H2O, 0.15% Na2O. MFCD00149140. Molecular Weight: 156.01. Molecular Formula: Al2O3∙3H2O or Al(OH)3. Color/form: white pwdr. Strem# 13-1000. http://www.strem.com/catalog/v/13-1000/
Catalog No. 50-901-09532