Learn More
Newcomer Supply Alpha Amylase 1%, Aqueous, 500 ml, Ready to-use glycogen digestion solution
SDP
Catalog No. NC1907206
Encompass
Newcomer Supply Alpha Amylase 1%, Aqueous is a convenient ready-to-use glycogen digestion solution for aiding in further identification of mucosubstances used in conjunction with the Periodic Acid Schiff (PAS) Stain procedure. Shelf Life is 18 months from date of manufacture. Store at 2-8°C.
Catalog No. NC1907206 Supplier Newcomer Supply Supplier No. 1905B
Newcomer Supply Alpha Amylase 1%, Aqueous is a convenient ready-to-use glycogen digestion solution for aiding in further identification of mucosubstances used in conjunction with the Periodic Acid Schiff (PAS) Stain procedure. Shelf Life is 18 months from date of manufacture. Store at 2-8°C.