Learn More
Ankom Technology ALPHA AMYLASE 250ML
SDP
Catalog No. NC9687428
Encompass
Alpha Amylase 250 ml. ANKOM FAA is a starch-hydrolyzing alpha-amylase enzyme with high heat and pH stability derived from a genetically modified strain of Bacillus licheniformis. Used for liquification of starch during Neutral Detergent Fiber Analysis.
Catalog No. NC9687428 Supplier Ankom Technology Supplier No. FAA
Alpha Amylase 250 ml. ANKOM FAA is a starch-hydrolyzing alpha-amylase enzyme with high heat and pH stability derived from a genetically modified strain of Bacillus licheniformis. Used for liquification of starch during Neutral Detergent Fiber Analysis.