Learn More
Sigma Aldrich Fine Chemicals Biosciences alpha-Amylase Bacillu250ML
Catalog No. 502903825
Encompass
Amylase belongs to glycosyl hydrolase family 13. It comprises of A B and C domains. Domain A has two aspartic acid and a glutamic acid as catalytic residues. Domain B possesses Ca2 and Na binding sites.
Catalog No. 50-290-3825 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A4862250ML
Amylase belongs to glycosyl hydrolase family 13. It comprises of A B and C domains. Domain A has two aspartic acid and a glutamic acid as catalytic residues. Domain B possesses Ca2 and Na binding sites.