Learn More
Sigma Aldrich Fine Chemicals Biosciences alpha-Amylase from Aspergi50G
Catalog No. 502901430
Encompass
Microbial amylases are exoenzymes and are used in several industrial applications such as production of bread maltose syrups and fermentation of soya sauce miso etc. Amylase isolated from Aspergillus oryzae has been found to have molecular weights of 51kDa (sedimentation and diffusion) and 49kDa (gel filtration).
Catalog No. 50-290-1430 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1006550G
Microbial amylases are exoenzymes and are used in several industrial applications such as production of bread maltose syrups and fermentation of soya sauce miso etc. Amylase isolated from Aspergillus oryzae has been found to have molecular weights of 51kDa (sedimentation and diffusion) and 49kDa (gel filtration).