Learn More
Sigma Aldrich Fine Chemicals Biosciences alpha Amylase from human s
Catalog No. 502903694
Encompass
amylase is the main secretory protein constituent of saliva. It contains 496 amino acids and has two isoforms glycosylated and non-glycosylated. amylase gene is located on human chromosome 1p21.1. amylase is mainly produced in pancreas and saliva.
Catalog No. 50-290-3694 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A05212.5KU
amylase is the main secretory protein constituent of saliva. It contains 496 amino acids and has two isoforms glycosylated and non-glycosylated. amylase gene is located on human chromosome 1p21.1. amylase is mainly produced in pancreas and saliva.