Learn More
Medchemexpress LLC Alpha-amylase, porcine pancreatic | 9000-90-2 | 100 KU
SDP
Catalog No. 5000410442
Encompass_Preferred
α-Amylase, Porcine Pancreatic (1,4-alpha-D-Glucan-glucanohydrolase) is a hydrolase enzyme isolated from porcine pancreatic. It catalyzes the hydrolysis of internal α-1, 4-glycosidic linkages in starch to yield products like glucose and maltose. This product is intended for life science research.
  • Can be used in the study of starch metabolism.
  • Shows binding activity toward N-linked oligosaccharides of glycoproteins, a unique feature not shared by α-amylases from saliva, wheat, or fungus.
  • Enhances the release of Dexamethasone from its γ-cyclodextrin complex at relatively low concentrations of γCD by catalyzing the hydrolysis of γCD.
Catalog No. 50-004-10442 Supplier Medchemexpress LLC Supplier No. HYB2193C100KU
α-Amylase, Porcine Pancreatic (1,4-alpha-D-Glucan-glucanohydrolase) is a hydrolase enzyme isolated from porcine pancreatic. It catalyzes the hydrolysis of internal α-1, 4-glycosidic linkages in starch to yield products like glucose and maltose. This product is intended for life science research.
  • Can be used in the study of starch metabolism.
  • Shows binding activity toward N-linked oligosaccharides of glycoproteins, a unique feature not shared by α-amylases from saliva, wheat, or fungus.
  • Enhances the release of Dexamethasone from its γ-cyclodextrin complex at relatively low concentrations of γCD by catalyzing the hydrolysis of γCD.