Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alpha-amylcinnamaldehyde (2-benzylideneheptanal) | 122-40-7 | MFCD00006988 | 99.6% | 202.29 g/mol | C14H18O | 50 G
SDP

Catalog No. 5000302967
Encompass_Preferred

α-Amylcinnamaldehyde (2-benzylideneheptanal, CAS 122-40-7) is a high-purity research chemical used as a ligand in studies of Niemann-Pick type C2 (NPC2) proteins and in fragrance and analytical research. Supplied for in vitro biochemical and pharmacological assays, it is suitable for protein-ligand interaction studies and analytical applications.

  • High purity (99.56%) suitable for in vitro studies.
  • Molecular formula C14H18O, molecular weight 202.29 g/mol.
  • Soluble in DMSO at ≥100 mg/mL for stock solutions.
  • Recommended storage: pure form -20°C for long term, 4°C for shorter term.
  • Useful as a ligand for NPC2 protein studies and as an aroma compound in flavor research.

Catalog No. 50-003-02967 Supplier Medchemexpress LLC Supplier No. HYW01478050G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

α-Amylcinnamaldehyde (2-benzylideneheptanal, CAS 122-40-7) is a high-purity research chemical used as a ligand in studies of Niemann-Pick type C2 (NPC2) proteins and in fragrance and analytical research. Supplied for in vitro biochemical and pharmacological assays, it is suitable for protein-ligand interaction studies and analytical applications.

  • High purity (99.56%) suitable for in vitro studies.
  • Molecular formula C14H18O, molecular weight 202.29 g/mol.
  • Soluble in DMSO at ≥100 mg/mL for stock solutions.
  • Recommended storage: pure form -20°C for long term, 4°C for shorter term.
  • Useful as a ligand for NPC2 protein studies and as an aroma compound in flavor research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.