Learn More
Medchemexpress LLC Alpha-Asarone | 2883-98-9 | 208.25 | 500 MG

Supplier: Medchemexpress LLC HYN0700500MG
Alpha-Asarone (α-Asarone) is one of the main psychoactive compounds and possesses antidepressant-like activity in mice. It is for research use only.
- Purity: 99.89%
- Appearance: Solid
- Color: White to off-white
- Initial source: Plants (Araceae, Acorus tatarinowii Schott)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.