Learn More
Medchemexpress LLC Alpha-boswellic acid | 471-66-9 | 456.70 | 1 ML
SDP
Catalog No. 5000388669
Encompass_Preferred
Alpha-Boswellic acid is an orally active pentacyclic triterpenoid compound extracted from frankincense. It exhibits anti-inflammatory and antitumor activities, making it suitable for Alzheimer's disease (AD) research.
  • Exhibits anti-inflammatory activity.
  • Shows antitumor activities.
  • Suitable for Alzheimer's disease (AD) research.
  • Reduces expression of pro-inflammatory cytokines.
  • Inhibits protein phosphorylations and reduces phosphorylated NF-κB protein levels.
  • Provides protective effects against ethanol-induced gastric injury.
  • Decreases epidermal thickening and inflammation in atopic-like dermatitis models.
Catalog No. 50-003-88669 Supplier Medchemexpress LLC Supplier No. HYN061110MM/1ML
Alpha-Boswellic acid is an orally active pentacyclic triterpenoid compound extracted from frankincense. It exhibits anti-inflammatory and antitumor activities, making it suitable for Alzheimer's disease (AD) research.
  • Exhibits anti-inflammatory activity.
  • Shows antitumor activities.
  • Suitable for Alzheimer's disease (AD) research.
  • Reduces expression of pro-inflammatory cytokines.
  • Inhibits protein phosphorylations and reduces phosphorylated NF-κB protein levels.
  • Provides protective effects against ethanol-induced gastric injury.
  • Decreases epidermal thickening and inflammation in atopic-like dermatitis models.