Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich alpha-Bromoisobutyryl bromide

Catalog No. 252271500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-227-1500G 500 g
25-227-1100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-227-1500G Supplier Sigma Aldrich Supplier No. 252271500G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • α-bromoisobutyryl bromide has been used:
  • As atom transfer radical polymerization (atrp) initiator for functionalization of hydroxyl groups present on the surface of graphene oxide
  • To form an n-protected halodienamide which provided four- and five-membered lactams in the presence of copper (i) and a tertiary amine
  • In preparation of polycaprolactone macroinitiator via reaction with oligomeric caprolactone diol and mesoporous silica nanoparticles with atrp initiator anchored on the exterior surface

Specifications

CAS 20769-85-1
Density 1.86 g/mL (at 25°C (literature))
Boiling Point 162°C to 164°C (lit.)
Molecular Formula C4H6Br2O
Linear Formula (CH3)2 CBrCOBr
Refractive Index n20/D 1.507 (literature)
MDL Number MFCD00000122
Quantity 500 g
Synonym 2-Bromo-2-methylpropionyl bromide; BIBB
Molecular Weight (g/mol) 229.9
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.