Learn More
Sigma Aldrich Fine Chemicals Biosciences alpha-Bungarotoxin, Bungarus multicinctus - CAS 11032-79-4 - Calbiochem Blocks neuromuscular transmission by irreversible binding to motor end-plate acetylcholine receptor (Kd = 1 pM to 1 nM) but does not depress acetylcholine release from motor nerve endings. | 11032-79-4 | 1MG
Catalog No. NC1565678
Encompass
alpha-Bungarotoxin, Bungarus multicinctus - CAS 11032-79-4 - Calbiochem Blocks neuromuscular transmission by irreversible binding to motor end-plate acetylcholine receptor (Kd = 1 pM to 1 nM) but does not depress acetylcholine release from motor nerve endings. | Mol Wt: 7984.12 | 11032-79-4 | 1MG
Catalog No. NC1565678 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2039801MG
alpha-Bungarotoxin, Bungarus multicinctus - CAS 11032-79-4 - Calbiochem Blocks neuromuscular transmission by irreversible binding to motor end-plate acetylcholine receptor (Kd = 1 pM to 1 nM) but does not depress acetylcholine release from motor nerve endings. | Mol Wt: 7984.12 | 11032-79-4 | 1MG