Learn More
CPC Scientific H-Arg-Tyr-Leu-Gly-Tyr-Leu-Glu-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific ACAS002A
Encompass
The alpha-casein was the most potent opioid in opioid activity bioassays.ONE-LETTER SEQUENCE: RYLGYLEMOLECULAR FORMULA: C43H64N10O12MOLECULAR WEIGHT:913.04STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [83471-49-2], RESEARCH AREA: PainREFERENCES: S.Loukas et al., Biochemistry, 22, 4567 (1983)
Catalog No. 50-210-8572