Learn More
Enzo Life Sciences alpha Catenin monoclonal antibody (15D9) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804101C100
Encompass_Preferred
Clone: 15D9. Isotype: IgG1. Immunogen: Recombinant human α-catenin. Species reactivity: Human, Mouse, Rat, Dog. Applications: ICC, IP, WB. Purity: Purified from serum-free cell culture supernatant by subsequent thiophilic adsorption and size exclusion chromatography. Formulation: Lyophilized from 1ml 2x PBS, 0.09% sodium azide, PEG and sucrose.
Catalog No. 50-200-9781