Learn More
Medchemexpress LLC Alpha-Cembrenediol | 57605-80-8 | 1 MG
SDP
Catalog No. 5000419707
Encompass_Preferred
Alpha-Cembrenediol is a potent inhibitor of EBV, inhibiting the early antigen of the Epstein-Bar virus. It also demonstrates anti-tumor activity. This product is for research use only and not sold to patients.
  • Potent inhibitor of Epstein-Barr virus (EBV)
  • Inhibits early antigen of Epstein-Barr virus
  • Demonstrates anti-tumor activity
  • Exhibits antiproliferative activity against various human cancer cell lines, including BT-474, MCF7, MDA-MB-231, MDA-MB-468, SK-BR-3, and T47D
  • Shows antimigratory activity against human MDA-MB-231 cells
  • Displays antiinvasive activity against human MDA-MB-231 cells
  • Possesses cytotoxicity against human MCF12A cells
Catalog No. 50-004-19707 Supplier Medchemexpress LLC Supplier No. HYN84591MG
Alpha-Cembrenediol is a potent inhibitor of EBV, inhibiting the early antigen of the Epstein-Bar virus. It also demonstrates anti-tumor activity. This product is for research use only and not sold to patients.
  • Potent inhibitor of Epstein-Barr virus (EBV)
  • Inhibits early antigen of Epstein-Barr virus
  • Demonstrates anti-tumor activity
  • Exhibits antiproliferative activity against various human cancer cell lines, including BT-474, MCF7, MDA-MB-231, MDA-MB-468, SK-BR-3, and T47D
  • Shows antimigratory activity against human MDA-MB-231 cells
  • Displays antiinvasive activity against human MDA-MB-231 cells
  • Possesses cytotoxicity against human MCF12A cells