Learn More
Medchemexpress LLC Alpha-chaconine | 20562-03-2 | MFCD09039102 | 99.8% | 852.06 g/mol | C45H73NO14 | 5 MG
SDP
Catalog No. 5000262525
Encompass_Preferred
Alpha-chaconine is a high-purity glycoalkaloid analytical standard (CAS 20562-03-2) supplied for research use. It serves as a reference material for analytical and biological studies and has been reported to inhibit COX-2 and iNOS expression, making it useful for inflammation-related research and assay development.
Catalog No. 50-002-62525 Supplier Medchemexpress LLC Supplier No. HY1291135MG
Alpha-chaconine is a high-purity glycoalkaloid analytical standard (CAS 20562-03-2) supplied for research use. It serves as a reference material for analytical and biological studies and has been reported to inhibit COX-2 and iNOS expression, making it useful for inflammation-related research and assay development.